Learn More
Sino Biological Recombinant Human CTHRC1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 11647H02H100
Encompass_Preferred
A DNA sequence encoding the Human CTHRC1 (NP_612464.1) (Met1-Lys243) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-241-2078