Learn More
ACROBiosystems Human CTLA-4 200ug
Supplier:  ACROBiosystems CT4H82E3200UG
Encompass_Preferred
Biotinylated Human CTLA-4 HisAvitag (CT4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession # P16410-1).
Catalog No. 50-253-5197