Learn More
ACROBiosystems ACROBiosystems Human CTLA-4 / CD152 Protein, Fc Tag
Supplier:  ACROBiosystems CT4H52551MG
Encompass_Preferred
Recombinant Protein;1MG;Human CTLA-4, Fc Tag (CT4-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # NP_005205.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.9 kDa. As a result of glycosylation, the protein migrates as 45-55 kDa under reducing (R) condition, and 90 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human B7-2, His Tag (Cat. No. CD6-H5223) at 10 μg/mL (100 μL/well) can bind Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) with a linear range of 2-78 ng/mL (QC tested).;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 10 μg/mL (100 μL/well) can bind Human B7-1, His Tag (Cat. No. B71-H5228) with a linear range of 0.156-1.25 μg/mL (Routinely tested).
Catalog No. 50-210-6615