Learn More
Sino Biological Recombinant Anti-CTLA-4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11159R06020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CTLA-4 / CD152 (rh CTLA-4 / CD152; Catalog#11159-H08H; NP_005205.2; Met1-Phe162).
Catalog No. 50-240-8148