Learn More
Sino Biological Anti-CTLA-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11159RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTLA4 extracellular domain (rh CTLA4; Catalog#11159-H08H; NP_005205.2; Met 1-Phe 162). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2413