Learn More
Sino Biological Anti-CTLA-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11159T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTLA-4 (rh CTLA-4; Catalog#11159-HNAH; NP_005205.2; Met1-Asp161). CTLA-4 specific IgG was purified by Human CTLA-4 affinity chromatography.
Catalog No. 50-255-4813