Learn More
Sino Biological Anti-CTLA-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11159T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CTLA-4 / CD152 (rh CTLA-4 / CD152; Catalog#11159-H08H; NP_005205.2; Met1-Phe162). CTLA-4 / CD152 specific IgG was purified by Human CTLA-4 / CD152 affinity chromatography.
Catalog No. 50-241-1092