Learn More
BIOHIPPO Human CTRB1 Microsample
SDP
Catalog No. 5000313800
Encompass
Human CTRB1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTRB1 in Serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 72 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CTRB1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CTRB1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CTRB1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13800 Supplier BIOHIPPO Supplier No. BHE15211548
Human CTRB1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTRB1 in Serum plasma and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 72 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CTRB1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CTRB1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CTRB1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color