Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CTRP4 Synthetic Peptide

Catalog No. PEP0270
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0270 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0270 Supplier Invitrogen™ Supplier No. PEP0270
Only null left
Add to Cart
Add to Cart

Synthetic Peptide

CTRP4 Synthetic Peptide, PEP-0270, from Invitrogen.

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of mouse CTRP4 have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 114900
Species Human
Purification Method purified
Accession Number Q9BXJ3
Common Name Human CTRP4
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol C1QTNF4
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.