Learn More
Sino Biological Anti-Cathepsin A Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10482MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CTSA (rh CTSA; Catalog#10482-H08H; NP_001121167.1; Met 1-Tyr 480). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8567