Learn More
Sino Biological Human CTSB PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10483RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cathepsin B / CTSB (rh Cathepsin B / CTSB; Catalog#10483-H08H; NP_001899.1; Arg18-Ile339). Cathepsin B / CTSB specific IgG was purified by Human Cathepsin B / CTSB affinity chromatography.
Catalog No. 50-237-2458