Learn More
Sino Biological Recombinant Anti-Cathepsin D Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12517R010100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin D / CTSD (rh Cathepsin D / CTSD; Catalog#12517-H08H; P07339; Met1-Leu412).
Catalog No. 50-255-4365