Learn More
Sino Biological Anti-Cathepsin S Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10487MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cathepsin S (rh Cathepsin S; Catalog#10487-H08H; NP_004070.3; Met1-Ile331). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7796