Learn More
BIOHIPPO Human CTSS Microsample
SDP
Catalog No. 5000315315
Encompass
Human CTSS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTSS in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CTSS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CTSS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CTSS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15315 Supplier BIOHIPPO Supplier No. BHE15211944
Human CTSS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTSS in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CTSS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CTSS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CTSS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color