Learn More
Biomatik Corporation Human CTXI (Cross Linked C-telopeptide of Type I Collagen) ELISA Kit
Supplier:  Biomatik Corporation EKF58565
Encompass_Preferred
Human CTXI (Cross Linked C-telopeptide of Type I Collagen) ELISA Kit for detecting CTXI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02452. Target Synonyms: CTXI; Cross Linked C-telopeptide of Type I Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9171