Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cu/ZnSOD ELISA Kit

Catalog No. 5018054
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-180-54 96 Tests
50-180-57 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-180-54 Supplier Invitrogen™ Supplier No. BMS222
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Cu/ZnSOD
HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
Human Cu/ZnSOD Standard, 5ng/mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Phosphate Buffered Saline Concentrate (PBS) 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

ELISA

The Human Cu/Zn Superoxide Dismutase(Hu Cu/ZnSOD)) ELISA quantitates Hu Cu/ZnSOD in human serum, plasma, amniotic fluid, urine, fetal umbilical vein blood, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Cu/ZnSOD. Principle of the method The Human Cu/ZnSOD solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00441
Assay Range 0.08-5.0 ng/mL
Assay Sensitivity 0.04 ng/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human Cu/ZnSOD
HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
Human Cu/ZnSOD Standard, 5 ng/mL
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Phosphate Buffered Saline Concentrate (PBS) 20x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Amniotic Fluid, Fetal Umbilical Vein Blood, Plasma, Serum, Supernatant, Urine
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodbALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodb; ALS; ALS1; HEL-S-44; IPOA; SOD; hSod1; homodimer
Gene ID (Entrez) 6647
Gene Symbol SOD1
Interassay CV 5.8%
Intraassay CV 5.1%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases Superoxide dismutase 1, hSod1
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Amniotic Fluid 10 μL, Fetal Umbilical Vein Blood 10 μL, Plasma 10 μL, Serum 10 μL, Supernatant 10 μL, Urine 10 μL
Storage Requirements 2-8°C
Synonym Cu/Zn Superoxide Dismutase
Target SOD1
Test Time 1 hr 20 min
Total Assay Time 1 hr 10 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.