Learn More
Biomatik Corporation Human CUB And Zona Pellucida Like Domains Protein 1 (CUZD1) ELISA Kit
Supplier:  Biomatik Corporation EKU03522
Encompass_Preferred
Human CUB And Zona Pellucida Like Domains Protein 1 (CUZD1) ELISA Kit for detecting CUZD1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86UP6. Gene ID: 50624. Target Synonyms: ERG-1; UO-44; Transmembrane protein UO-44. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4733