Learn More
Biomatik Corporation Human CUB Domain Containing Protein 1 (CDCP1) CLIA Kit
Supplier:  Biomatik Corporation EKU09925
Encompass_Preferred
Human CUB Domain Containing Protein 1 (CDCP1) CLIA Kit for detecting CDCP1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9H5V8. Gene ID: 64866. Target Synonyms: CD318; CDCP1; SIMA135; TRASK; Membrane glycoprotein gp140; Transmembrane and associated with src kinases; Subtractive immunization M plus HEp3-associated 135 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6911