Learn More
Sino Biological Recombinant Anti-Fractalkine/CX3CL1 Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10636R409H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fractalkine / CX3CL1 (rh Fractalkine / CX3CL1; Catalog#10636-H08H; NP_002987.1; Met 1-Arg 339), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-0480