Learn More
Sino Biological Human CX3CL1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10636R409100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fractalkine / CX3CL1 (rh Fractalkine / CX3CL1; Catalog#10636-H08H; NP_002987.1; Met 1-Arg 339).
Catalog No. 50-237-3868