Learn More
BIOHIPPO Human CX3CL1 Microsample
SDP
Catalog No. 5000314450
Encompass
Human CX3CL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CX3CL1 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CX3CL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CX3CL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CX3CL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14450 Supplier BIOHIPPO Supplier No. BHE15212239
Human CX3CL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CX3CL1 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CX3CL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CX3CL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CX3CL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color