Learn More
Sino Biological Recombinant Human Cx3cr1 Protein (VLP, Flag & Strep Tag)
Catalog No. 502993614
Encompass_Preferred
A DNA sequence encoding the Human CX3CR1-VLP (P49238) (Met1-Leu315,thermostabilizing mutations) was expressed, with a Flag tag at the N-terminus, and a strep tag at the C-terminus.
Catalog No. 50-299-3614 Supplier Sino Biological Supplier No. 12505H84H20
A DNA sequence encoding the Human CX3CR1-VLP (P49238) (Met1-Leu315,thermostabilizing mutations) was expressed, with a Flag tag at the N-terminus, and a strep tag at the C-terminus.