Learn More
Sino Biological Human CXADR/CAR Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785975
Encompass_Preferred
The Human CXADR / CAR ELISA Pair Set is for the quantitative determination of Human CXADR / CAR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CXADR / CAR coated on a 96-well plate. Standards and samples are added to the wells, and any Human CXADR / CAR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human CXADR / CAR monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CXADR / CAR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85975 Supplier Sino Biological Supplier No. SEK1079915
The Human CXADR / CAR ELISA Pair Set is for the quantitative determination of Human CXADR / CAR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CXADR / CAR coated on a 96-well plate. Standards and samples are added to the wells, and any Human CXADR / CAR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human CXADR / CAR monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CXADR / CAR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.