Learn More
Sino Biological Human CXADR MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10799R271100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR / CAR (rh CXADR / CAR; Catalog#10799-H08H; NP_001329.1; Met1-Gly237).
Catalog No. 50-237-1944