Learn More
Sino Biological Recombinant Anti-CXADR/CAR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10799R02320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR / CAR (rh CXADR / CAR; Catalog#10799-H08H; NP_001329.1; Met1-Gly237).
Catalog No. 50-241-2418