Learn More
Sino Biological Human CXCL1 ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological 10202MM0220
Encompass_Preferred
Each vial contains 18 ng of recombinant human CXCL1. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 200 pg/mL is recommended.
Catalog No. 50-237-4384