Learn More
Sino Biological Recombinant Anti-CXCL1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10877R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXCL1 (rh CXCL1; Catalog#10877-HNCE; P09341-1; Ala35-Asn107).
Catalog No. 50-255-5118