Learn More
Sino Biological Human CXCL1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10877MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CXCL1 (rh CXCL1; Catalog#10877-HNCE; P09341-1; Ala35-Asn107). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2880