Learn More
Invitrogen™ Human CXCL10 (IP-10) Recombinant Protein
Description
RP-75644 contains purified recombinant human IFN Inducible Protein protein with an N-terminal hexahistidine tag. RP-75644 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P02778 |
| Concentration | 0.30 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 3627 |
| Name | Human CXCL10 (IP-10) |
| pH Range | 8.0 |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.