Learn More
Sino Biological Human CXCL11 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10876MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human I-TAC / CXCL11 (rh I-TAC / CXCL11; Catalog#10876-HNAE; O14625; Phe22-Phe94). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3424