Learn More
Sino Biological Human CXCL11 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785734
Encompass_Preferred
The Human I-TAC / CXCL11 ELISA Pair Set is for the quantitative determination of Human I-TAC / CXCL11.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human I-TAC / CXCL11 coated on a 96-well plate. Standards and samples are added to the wells, and any Human I-TAC / CXCL11 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human I-TAC / CXCL11 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human I-TAC / CXCL11 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85734 Supplier Sino Biological Supplier No. SEK1087615
The Human I-TAC / CXCL11 ELISA Pair Set is for the quantitative determination of Human I-TAC / CXCL11.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human I-TAC / CXCL11 coated on a 96-well plate. Standards and samples are added to the wells, and any Human I-TAC / CXCL11 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human I-TAC / CXCL11 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human I-TAC / CXCL11 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.