Learn More
Sino Biological Anti-CXCL11 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10876RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human I-TAC / CXCL11 (rh CXCL11; Catalog#10876-HNAE; O14625; Phe22-Phe94). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0471