Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human CXCL12 (SDF-1g) Recombinant Protein, PeproTech®

Catalog No. 502387944
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-238-7944 10 μg
50-238-7937 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-238-7944 Supplier Gibco™ Supplier No. 30028G10UG
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human CXCL12 (SDF-1g) is a 11.6 kDa protein containing 98 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The SDF-1 proteins are stromal-derived CXC chemokines that signal primarily through the CXCR4 receptor. SDF-1 proteins exert chemotactic activity over a broad range of immune system cell types and may also be involved in other processes including metastasis of certain cancers, inflammation, and angiogenesis. Additionally, SDF-1 can suppress HIV entry into cells expressing the CXCR4 receptor. Five isomers of SDF-1 have been found with SDF-1alpha being the predominant form, expressing in a wide range of cell types. The SDF-1gamma isoform is distinctive in that it contains a highly basic C-terminal region, which presumably is the reason for its high binding affinity to gylcosaminoglycans (GAG) and heparin sulfate (HS). Consequently, most of SDF-1gamma exists in a cell- and membrane-bound form, which confers greater stability compared to other isoforms via increased resistance to proteolytic degradation. SDF-1gamma binds with reduced affinity to the CXCR4 receptor when compared to the alpha and beta isoforms, but because of its increased stability can, over time, exert equal or greater activity when compared to other SDF-1 isoforms. SDF-1gamma expression has been found primarily in the heart (human and mouse) and in the brain and central nervous system (rat).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48061
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 6387
Molecular Weight (g/mol) 11.6 kDa
Name Human CXCL12 (SDF-1 g)
Purification Method HPLC, SDS-PAGE
Quantity 10 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias PBSF; Stromal-Cell Derived Factor-1
Common Name CXCL12 gamma
Gene Symbol CXCL12
Biological Activity Determined by its ability to chemoattract human T cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKGR REEKVGKKEK IGKKKRQKKR KAAQKRKN
Content And Storage -20°C
Activity Determined by its ability to chemoattract human T cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.