Learn More
Sino Biological Recombinant Human SDF-1/CXCL12 Protein (isoform a)
Supplier:  Sino Biological 13511HNCE100
Encompass_Preferred
A DNA sequence encoding the human CXCL12 isoform alpha (P48061-2) (Lys 22-Lys 89) was expressed and purified.
Catalog No. 50-224-6729