Learn More
Sino Biological Recombinant Human SDF-1/CXCL12 Protein (isoform a, His Tag)
Supplier:  Sino Biological 13511H07E50
Encompass_Preferred
A DNA sequence encoding the human CXCL12 isoform alpha (P48061-2) (Lys 22-Lys 89) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6797