Learn More
Sino Biological Recombinant Human SDF-1/CXCL12 Protein (hFc Tag)
Supplier:  Sino Biological 10118H01H50
Encompass_Preferred
A DNA sequence encoding the N-terminally truncated human SDF1β (NP_000600.1) (Lys 22-Met 93) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-7037