Learn More
Bon Opus Biosciences Recombinant Human CXCL13 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP104050UG
Encompass_Preferred
Recombinant Human CXCL13 (C-hFc) is produced by Human Cells. The target gene encoding Val23-Pro109 is expressed with a C-hFc tag. Accession# O43927. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1040
Catalog No. 50-217-2639