Learn More
Sino Biological Recombinant Human BRAK/CXCL14 Protein 1 mg
Supplier:  Sino Biological 10880HNAE1
Encompass_Preferred
A DNA sequence encoding human CXCL14 (O95715) (Ser35-Glu111) was expressed with a N-terminal Met.
Catalog No. 50-224-7041