Learn More
Sino Biological Human CXCL14 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10880H07E120
Encompass_Preferred
A DNA sequence encoding the human CXCL14 (O95715) (Ser35-Glu111) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2643