Learn More
Sino Biological Anti-GRO Beta/CXCL2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10586MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CXCL2/MIP-2 (rh CXCL2/MIP-2; Catalog#10586-HNCE; NP_002080.1; Ala35-Asn107). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1317