Learn More
Abcam Human CXCL3 ELISA Kit, assay type: Sandwich ELISA, sample type: serum, sensitivity: 1.1 pg/mL, detection range: 1.56 - 100 pg/mL, assay time: 1h 30m, colorimetric detection
Supplier: Abcam AB2345741X96TESTS
The Abcam Human CXCL3 ELISA Kit is a highly sensitive immunoassay designed for the quantitative detection of human CXCL3 in serum samples. This kit features a streamlined SimpleStep Sandwich ELISA protocol that reduces overall assay time, making it efficient and user-friendly. It provides a reliable method for quantifying CXCL3, which plays crucial roles in various biological processes.
- Assay type: Sandwich ELISA (quantitative)
- Assay platform: Pre-coated microplate (12 x 8 well strips)
- Detection method: Colorimetric
- Sample types: Serum
- Reacts with: Human
- Target name: Human CXCL3 (GRO3, MIP2b)
- Sensitivity: 1.1 pg/mL
- Detection range: 1.56 - 100 pg/mL
- Assay time: 1 h 30 m
- Assay protocol: Single-wash, single incubation
- Protocol steps: Mix, wash, read
- Reproducibility: Consistent results in human serum
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.