Learn More
Sino Biological Anti-CXCL5 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10889MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CXCL5 (rh CXCL5; Catalog#10889-HNAE; P42830; Arg45-Asn114). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8022