Learn More
Sino Biological Anti-CXCL5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10889T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CXCL5 (rh CXCL5; Catalog#10889-HNAE; P42830; Arg45-Asn114). CXCL5 specific IgG was purified by Human CXCL5 affinity chromatography.
Catalog No. 50-241-1211