Learn More
Kingfisher Biotech Inc Human GM-CSF ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1996H003
Encompass_Preferred
The Human GM-CSF ELISA contains capture antibody, standard, and detection antibody for development of a Human GM-CSF ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CSF2, GMCSF, colony stimulating factor 2, CSF. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5653