Learn More
Sino Biological Human CXCL8 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370835
Encompass_Preferred
Each vial contains 3 ng of recombinant Human IL-8/Interleukin-8/CXCL8. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 240 pg/mL is recommended.
Catalog No. 50-237-0835 Supplier Sino Biological Supplier No. SEK1009815
Each vial contains 3 ng of recombinant Human IL-8/Interleukin-8/CXCL8. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 240 pg/mL is recommended.