Learn More
Sino Biological Human CXCL8 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11039R014100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-8/Interleukin-8/CXCL8 (rh IL-8/Interleukin-8/CXCL8; Catalog#10098-H08Y; NP_000575.1; Ser28-Ser99). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4417