Learn More
Sino Biological Human CXCL9 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502372842
Encompass_Preferred
Each vial contains 13 ng of recombinant Human CXCL9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 600 pg/mL is recommended.
Catalog No. 50-237-2842 Supplier Sino Biological Supplier No. SEK108885
Each vial contains 13 ng of recombinant Human CXCL9. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 600 pg/mL is recommended.