Learn More
Proteintech Group Inc Human CXCL9/MIG ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE0016596T
Encompass_Preferred
CXCL9, also named MIG, is a member of the CXC family and has an important role in the chemotaxis of immune cells. CXCL9 is secreted by various cell types including immune cells (T lymphocytes, NK cells, dendritic cells, macrophages, eosinophils etc.), and non-immune cells (hepatic stellate cells, preadipocytes, thyrocytes, endothelial cell, tumor cells, and fibroblasts etc). CXCL9 plays role to induce chemotaxis, promote differentiation and multiplication of leukocytes, and cause tissue extravasation. Functions of CXCL9 are mostly mediated through its receptor CXCR3, which initiates intracellular signaling pathways like p38 and Erk. CXCL9 is involved inthe pathogenesis of a variety of physiologic diseases during their initiation andtheir maintenance.
Catalog No. 50-197-4120