Learn More
Sino Biological Anti-CXCL9/MIG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10888RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CXCL9 / MIG (rh CXCL9 / MIG; Catalog#10888-HNAE; Q07325; Thr 23-Thr 125). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1226