Learn More
Sino Biological Recombinant Human CXCL9/MIG Protein
Supplier:  Sino Biological 10888HNAE1
Encompass_Preferred
A DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125) was expressed and purified.
Catalog No. 50-255-5099